Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GeminiBio™ GemCell™ Plus Xeno-Free, World Grade
SDP

Catalog No. 11852110
Change view
Click to view available options
Sterilization Method:
Gamma-irradiated
Sequential-filtered, 100 nm
Format:
Bag - BioProcess Container (BPC)
Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterilization Method Format
11-852-110 Sequential-filtered, 100 nm Bag - BioProcess Container (BPC)
11-852-109 Sequential-filtered, 100 nm Bottle
11-852-104 Gamma-irradiated Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
For further manufacturing or research use only. Not for direct Therapeutic use.
Catalog No. 11-852-110 Supplier GeminiBio™ Supplier No. 100916750
Add to Cart
Add to Cart
For further manufacturing or research use only. Not for direct Therapeutic use.

Collected from a controlled pool of up to 16 healthy male AB donors and defibrinated using therapeutic-grade recombinant human thrombin. Xeno-Free and extensively tested to meet EU requirements for clinical and commercial use.

GemCell™ Plus Xeno-Free, World Grade is designed for global programs requiring tighter donor control and enhanced traceability. A capped donor pool and Xeno-Free processing support alignment with EU-focused regulatory strategies and region-specific qualification requirements.

  • Collected from a controlled pool of up to 16 healthy male AB donors
  • Defibrinated using therapeutic-grade recombinant human thrombin
  • Manufactured through a Xeno-Free process
  • Extensively tested to support clinical and commercial use
  • Controlled donor strategy enhances traceability and lot consistency

Suited for:

  • Global or EU-aligned programs requiring enhanced donor control
  • Applications prioritizing traceability and consistency
  • Customers balancing regulatory alignment with defined supply strategies

Specifications

Form Liquid
Format Bag - BioProcess Container (BPC)
Product Type Human Serum
Quantity 100 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.