Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GeminiBio™ GemCell™ Plus Xeno-Free
SDP

Catalog No. 11852111
Change view
Click to view available options
Sterilization Method:
Gamma-irradiated
Sequential-filtered, 100 nm
Format:
Bag - BioProcess Container (BPC)
Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterilization Method Format
11-852-111 Sequential-filtered, 100 nm Bottle
11-852-103 Sequential-filtered, 100 nm Bag - BioProcess Container (BPC)
11-852-108 Gamma-irradiated Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
For further manufacturing or research use only. Not for direct Therapeutic use.
Catalog No. 11-852-111 Supplier GeminiBio™ Supplier No. 100912100
Add to Cart
Add to Cart
For further manufacturing or research use only. Not for direct Therapeutic use.

Collected from healthy male AB donors and defibrinated using therapeutic-grade recombinant human thrombin. Manufactured Xeno-Free and extensively tested to support clinical and commercial applications with scalability enabled by a large donor pool.

GemCell™ Plus Xeno-Free is designed for cell therapy programs advancing beyond development into clinical and commercial manufacturing. Built on a Xeno-Free manufacturing process and supported by a large donor pool, this grade helps balance regulatory alignment with long-term supply continuity.

  • Collected from healthy male donors of the AB serotype
  • Defibrinated using therapeutic-grade recombinant human thrombin
  • Manufactured through a Xeno-Free process
  • Extensively tested to support clinical and commercial applications
  • Large donor pool enables scalability and long-term supply planning

Suited for:

  • Programs transitioning from development into clinical or commercial manufacturing
  • Applications prioritizing Xeno-Free processing
  • Customers requiring scalable donor strategies and supply continuity

Specifications

Form Liquid
Format Bottle
Product Type Human Serum
Quantity 100 mL
Sterility Sterile-filtered
Sterilization Method Sequential-filtered, 100 nm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.