Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GeminiBio GemCell™ U.S. Origin Fetal Bovine Serum
SDP

Catalog No. 11852139 Shop All Gemini Bio Products
Change view
Click to view available options
Gamma-irradiated:
No
Yes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gamma-irradiated
11-852-139 Yes
11-852-138 No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-852-139 Supplier GeminiBio Supplier No. 100500G500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fully traceable serum that supports growth of a wide range of cell types while minimizing experiment-to-experiment variability.

  • Sourced exclusively from the United States for full origin traceability
  • Meets stringent industry standards with low endotoxin and hemoglobin levels
  • Triple 0.1 μm sterile-filtered for exceptional purity
  • Mycoplasma-tested and virus-screened
  • Supports growth of a wide range of cell types while minimizing experiment-to-experiment variability
  • Available standard or gamma-irradiated
  • Up to 5-year shelf life from date of manufacture
  • Ships on dry ice; store frozen at -20°C to -10°C
  • Ideal for general cell culture, stem cell research, immunology, neuroscience, cancer research, and biotechnology
  • Manufactured under an ISO 13485:2016 certified quality system
  • Complimentary heat inactivation upon request

Specifications

Additional Assays Biochemical profile, Electrophoretic profile
Certifications/Compliance ISO 13485:2016 certified quality system
Content And Storage 500 mL bottle, store frozen at -20°C to -10°C
Species Bovine
Age Fetal
Country of Origin United States
Form Liquid
Format Bottle
Product Type Fetal Bovine Serum
Quantity 500 mL
Serum Treatment Gamma-irradiated
Sterility Sterile
Sterilization Method Triple-filtered, 0.1 μm, Gamma-irradiated
Source Fetal bovine blood
Grade Premium
Processing Triple 0.1 μm sterile-filtered, gamma-irradiated
Tested For Mycoplasma, Sterility (bacteria and fungi), Adventitious viruses
Formulation Liquid, frozen
For Use With (Application) Cell Culture
Heat Inactivated No
Lyophilized No
Product Line GemCell™
Mycoplasma Detected No
Shipping Condition Dry Ice
Regulatory Status RUO
Gamma-irradiated Yes
Show More Show Less

For cell culture, further manufacturing or research use only. Not for direct therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.