Learn More
Bioworld GemCell U.S. Origin Human Serum AB, 100 mL
Catalog No. 50254210
Encompass_Preferred
GemCell™ human serum AB is collected from healthy male donors of the AB serotype at FDA-licensed facilities in the United States. This material is defibrinated from Source Plasma AB. All donor units are tested for viral markers (HIVAg, HBsAg, HIV 1 and 2) and found to be nonreactive.Specifications0.1µm sterile-filteredShips on dry ice.
Catalog No. 50-254-210 Supplier Bioworld Supplier No. 306110921
GemCell™ human serum AB is collected from healthy male donors of the AB serotype at FDA-licensed facilities in the United States. This material is defibrinated from Source Plasma AB. All donor units are tested for viral markers (HIVAg, HBsAg, HIV 1 and 2) and found to be nonreactive.Specifications0.1µm sterile-filteredShips on dry ice.