Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GeminiBio™ GemCell™ U.S. Origin Human Serum AB
SDP

Catalog No. 11852100 Shop All Gemini Bio Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-852-100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
For further manufacturing or research use only. Not for direct Therapeutic use.
Catalog No. 11-852-100 Supplier GeminiBio™ Supplier No. 100512G100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For further manufacturing or research use only. Not for direct Therapeutic use.

Collected from healthy male AB donors and defibrinated from source plasma using recombinant human thrombin. Designed for R and D use, it provides consistent performance and lot reliability supported by a large U.S. donor pool.

GemCell™ (US Origin) is designed to support research and early development programs that require consistency, availability, and defined processing. Produced from healthy male AB donors and defibrinated using recombinant human thrombin, GemCell™ delivers a reliable Human Serum AB option for R and D workflows.

  • Collected from healthy male donors of the AB serotype
  • Defibrinated from source plasma using recombinant human thrombin
  • Designed specifically for research and development applications
  • Large donor pool supports lot availability and continuity during early-stage programs
  • Defined processing approach supports consistency across R and D workflows

Suited for:

  • Discovery and R and D programs requiring reliable access and consistency
  • Early-stage workflows where donor scale and availability are priorities
  • Teams establishing foundational data ahead of clinical translation

Specifications

Form Liquid
Format Bottle
Product Type Human Serum
Quantity 100 mL
Sterility Sterile-filtered
Sterilization Method Sequential-filtered, 100 nm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.