Learn More
Cayman Chemical Gemcitabine-13C 15N2 hydroch
SDP
Catalog No. 502986771
Encompass_Preferred
An internal standard for the quantification of gemcitabine by GC- or LC-MS
Catalog No. 50-298-6771 Supplier Cayman Chemical Supplier No. 22560500 UG
An internal standard for the quantification of gemcitabine by GC- or LC-MS