Learn More
Selleck Chemical LLC Gemcitabine S1714-200mg
SDP
Catalog No. 503108139
Encompass_Preferred
Gemcitabine a nucleic acid synthesis inhibitor is a very potent and specific deoxycytidine analogue used as chemotherapy Gemcitabine induces a potent p53-dependent apoptosis
Catalog No. 50-310-8139 Supplier Selleck Chemical LLC Supplier No. S1714200MG
Gemcitabine a nucleic acid synthesis inhibitor is a very potent and specific deoxycytidine analogue used as chemotherapy Gemcitabine induces a potent p53-dependent apoptosis