Learn More
Abcam Gemcitabine, Cytidine analog, 100MG
Catalog No. 502705081
Encompass_Preferred
MW 263.2 Da, Purity >98%. Cytidine analog. Antimetabolite. Induces apoptosis. Blocks cytosine nucleotide conversion into the deoxy derivative and blocks incorporation of the thymidine nucleotide into the DNA strand. Shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5081 Supplier Abcam Supplier No. AB145657100MG
MW 263.2 Da, Purity >98%. Cytidine analog. Antimetabolite. Induces apoptosis. Blocks cytosine nucleotide conversion into the deoxy derivative and blocks incorporation of the thymidine nucleotide into the DNA strand. Shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement