Learn More
Medchemexpress LLC Gemcitabine elaidate | 210829-30-4 | MFCD15145214 | 98.9% | C27H43F2N3O5 | 10MG
SDP
Catalog No. 5000259962
Encompass_Preferred
Gemcitabine elaidate is a lipophilic prodrug of gemcitabine intended to improve membrane permeability and delivery of the active nucleoside. It is enzymatically cleaved by esterases to release gemcitabine and is used as a research reagent in preclinical in vitro and in vivo tumor models.
  • Lipophilic prodrug that enhances membrane permeability
  • Converted to gemcitabine by esterases in biological systems
  • Used in preclinical anti-tumor in vitro and in vivo studies
  • High purity small-molecule suitable for analytical and biological assays
  • Supplied as a powdered research reagent for laboratory use
Catalog No. 50-002-59962 Supplier Medchemexpress LLC Supplier No. HY1353810MG
Gemcitabine elaidate is a lipophilic prodrug of gemcitabine intended to improve membrane permeability and delivery of the active nucleoside. It is enzymatically cleaved by esterases to release gemcitabine and is used as a research reagent in preclinical in vitro and in vivo tumor models.
  • Lipophilic prodrug that enhances membrane permeability
  • Converted to gemcitabine by esterases in biological systems
  • Used in preclinical anti-tumor in vitro and in vivo studies
  • High purity small-molecule suitable for analytical and biological assays
  • Supplied as a powdered research reagent for laboratory use