Learn More
Medchemexpress LLC Gemcitabine elaidate hydrochloride | 2918768-08-6 | 99.9% | 564.11 g/mol | C27H44ClF2N3O5 | 5 MG
SDP
Catalog No. 5000223306
Encompass_Preferred
Gemcitabine elaidate hydrochloride is a lipophilic prodrug of gemcitabine provided for research use. It is enzymatically converted by esterases to gemcitabine, which can be phosphorylated intracellularly and exhibits antitumor activity in preclinical models. The material is supplied as a high-purity solid for laboratory applications and handling under appropriate safety conditions is required.
Catalog No. 50-002-23306 Supplier Medchemexpress LLC Supplier No. HY13538A5MG
Gemcitabine elaidate hydrochloride is a lipophilic prodrug of gemcitabine provided for research use. It is enzymatically converted by esterases to gemcitabine, which can be phosphorylated intracellularly and exhibits antitumor activity in preclinical models. The material is supplied as a high-purity solid for laboratory applications and handling under appropriate safety conditions is required.