Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gemcitabine elaidate hydrochloride | 2918768-08-6 | 99.9% | C27H44ClF2N3O5 | 10MG
SDP

Catalog No. 5000259894
Encompass_Preferred

Gemcitabine elaidate hydrochloride is a lipophilic prodrug of gemcitabine that is enzymatically converted to the active nucleoside analogue by esterases and has reported antitumor activity in preclinical studies. It is supplied as a research-grade solid for laboratory use.

  • Lipophilic prodrug designed for improved cellular uptake.
  • Enzymatically converted to active gemcitabine by esterases.
  • Reported antitumor activity in preclinical models.
  • High chemical purity: 99.9%.
  • Chemical formula C27H44ClF2N3O5; molecular weight 564.11 g·mol⁻¹.
  • Supplied in small research-grade pack sizes suitable for in vitro work.

Catalog No. 50-002-59894 Supplier Medchemexpress LLC Supplier No. HY13538A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gemcitabine elaidate hydrochloride is a lipophilic prodrug of gemcitabine that is enzymatically converted to the active nucleoside analogue by esterases and has reported antitumor activity in preclinical studies. It is supplied as a research-grade solid for laboratory use.

  • Lipophilic prodrug designed for improved cellular uptake.
  • Enzymatically converted to active gemcitabine by esterases.
  • Reported antitumor activity in preclinical models.
  • High chemical purity: 99.9%.
  • Chemical formula C27H44ClF2N3O5; molecular weight 564.11 g·mol⁻¹.
  • Supplied in small research-grade pack sizes suitable for in vitro work.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.