Learn More
Selleck Chemical LLC Gemcitabine 50mg 95058-81-4 LY-188011, NSC 613327
SDP
Catalog No. NC0476353
Encompass_Preferred
Gemcitabine (LY-188011, NSC 613327), a nucleic acid synthesis inhibitor, is a very potent and specific deoxycytidine analogue, used as chemotherapy. Gemcitabine induces a potent p53-dependent apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0476353 Supplier Selleck Chemical LLC Supplier No. S171450MG
Gemcitabine (LY-188011, NSC 613327), a nucleic acid synthesis inhibitor, is a very potent and specific deoxycytidine analogue, used as chemotherapy. Gemcitabine induces a potent p53-dependent apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.