Learn More
Enzo Life Sciences Gemcitabine . hydrochloride (100mg). CAS: 122111-03-9
SDP
Supplier:  Enzo Life Sciences ALX480101M100
Encompass_Preferred
Deoxycytidine analog with cytotoxic effects. Induces apoptosis and inhibits DNA synthesis. Alternative name: 2'-Deoxy-2',2'-difluorocytidine . HCl, LY-188011 . HCl. Purity: ≥97.5% (Assay). Solubility: Soluble in water; slightly soluble in methanol; almost insoluble in 100% ethanol or polar organic solvents. Long Term Storage: +4°C.
Catalog No. 50-200-9333