Learn More
Selleck Chemical LLC Gemcitabine HCl S1149-10g
SDP
Catalog No. 503108426
Encompass_Preferred
Gemcitabine HCl is a DNA synthesis inhibitor with IC50 of 50 nM 40 nM 18 nM and 12 nM in PANC1 MIAPaCa2 BxPC3 and Capan2 cells respectively
Catalog No. 50-310-8426 Supplier Selleck Chemical LLC Supplier No. S114910G
Gemcitabine HCl is a DNA synthesis inhibitor with IC50 of 50 nM 40 nM 18 nM and 12 nM in PANC1 MIAPaCa2 BxPC3 and Capan2 cells respectively