Learn More
Medchemexpress LLC HY-B0003 200mg , Gemcitabine (hydrochloride) CAS:122111-03-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0003200MG
Encompass_Preferred
HY-B0003 200mg Gemcitabine (hydrochloride) CAS: 122111-03-9 Gemcitabine Hydrochloride (LY 188011 Hydrochloride) is a pyrimidine nucleoside analog antimetabolite and an antineoplastic agent. Gemcitabine Hydrochloride inhibits DNA synthesis and repair, resulting in autophagyand apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9571