Learn More
Medchemexpress LLC Gemcitabine Hydrochloride | 122111-03-9 | 99.9% | C9H11F2N3O4 · HCl | 5 G
SDP
Catalog No. 5000432935
Encompass_Preferred
Gemcitabine Hydrochloride (LY 188011 Hydrochloride) is a pyrimidine nucleoside analog antimetabolite and an antineoplastic agent. It works by inhibiting DNA synthesis and repair, ultimately leading to autophagy and apoptosis.
  • Targets DNA synthesis.
  • Inhibits the growth of various cancer cell lines, including BxPC-3, Mia Paca-2, PANC-1, PL-45, and AsPC-1.
  • Effectively kills both mouse and human senescent cells in vitro.
  • Demonstrates increased survival time in mouse models when combined with DMAPT.
  • Can be administered via endotracheal spray in rats with lower toxicity compared to oral administration at certain dosages.
Catalog No. 50-004-32935 Supplier Medchemexpress LLC Supplier No. HYB00035G
Gemcitabine Hydrochloride (LY 188011 Hydrochloride) is a pyrimidine nucleoside analog antimetabolite and an antineoplastic agent. It works by inhibiting DNA synthesis and repair, ultimately leading to autophagy and apoptosis.
  • Targets DNA synthesis.
  • Inhibits the growth of various cancer cell lines, including BxPC-3, Mia Paca-2, PANC-1, PL-45, and AsPC-1.
  • Effectively kills both mouse and human senescent cells in vitro.
  • Demonstrates increased survival time in mouse models when combined with DMAPT.
  • Can be administered via endotracheal spray in rats with lower toxicity compared to oral administration at certain dosages.