Learn More
Medchemexpress LLC Gemcitabine hydrochloride | 122111-03-9 | 99.9% | C9H12ClF2N3O4 | 25 MG
SDP
Catalog No. 5000357970
Encompass_Preferred
Gemcitabine hydrochloride is a pyrimidine nucleoside analog antimetabolite and an antineoplastic agent. It functions by inhibiting DNA synthesis and repair, leading to autophagy and apoptosis, and is primarily used in cancer research. It appears as a white to off-white solid.
  • Inhibits DNA synthesis and repair
  • Induces autophagy and apoptosis
  • Targets DNA/RNA synthesis
  • Functions as a nucleoside antimetabolite/analog
  • Appears as a white to off-white solid
Catalog No. 50-003-57970 Supplier Medchemexpress LLC Supplier No. 25MGHYB0003R
Gemcitabine hydrochloride is a pyrimidine nucleoside analog antimetabolite and an antineoplastic agent. It functions by inhibiting DNA synthesis and repair, leading to autophagy and apoptosis, and is primarily used in cancer research. It appears as a white to off-white solid.
  • Inhibits DNA synthesis and repair
  • Induces autophagy and apoptosis
  • Targets DNA/RNA synthesis
  • Functions as a nucleoside antimetabolite/analog
  • Appears as a white to off-white solid