Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gemcitabine monophosphate | 116371-67-6 | 100.0% | 343.18 | C9H12F2N3O7P | 5 MG
SDP

Catalog No. 5000264568
Encompass_Preferred

Gemcitabine monophosphate (gemcitabine 5′-phosphate) is the 5′-phosphate intermediate of gemcitabine supplied as a solid for in vitro research. It is used as an active metabolite/intermediate in biochemical and anti-cancer studies and for formulation research such as nanoparticle delivery. For research use only.

  • CAS number: 116371-67-6
  • Molecular formula: C9H12F2N3O7P
  • Molecular weight: 343.18
  • Purity: 99.98% (manufacturer stated)
  • Appearance: white to off-white solid
  • Solubility: DMSO 200 mg/mL, water 100 mg/mL (both may require ultrasonic)
  • Storage: sealed, away from moisture; in solvent -80°C up to 6 months
  • Available pack sizes: small mg quantities (e.g., 5 mg) and larger sizes on request

Catalog No. 50-002-64568 Supplier Medchemexpress LLC Supplier No. HY1089325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gemcitabine monophosphate (gemcitabine 5′-phosphate) is the 5′-phosphate intermediate of gemcitabine supplied as a solid for in vitro research. It is used as an active metabolite/intermediate in biochemical and anti-cancer studies and for formulation research such as nanoparticle delivery. For research use only.

  • CAS number: 116371-67-6
  • Molecular formula: C9H12F2N3O7P
  • Molecular weight: 343.18
  • Purity: 99.98% (manufacturer stated)
  • Appearance: white to off-white solid
  • Solubility: DMSO 200 mg/mL, water 100 mg/mL (both may require ultrasonic)
  • Storage: sealed, away from moisture; in solvent -80°C up to 6 months
  • Available pack sizes: small mg quantities (e.g., 5 mg) and larger sizes on request

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.