Learn More
Medchemexpress LLC Gemcitabine monophosphate | 116371-67-6 | 99.98% | 343.18 | 10 MG
SDP
Catalog No. 5000345774
Encompass_Preferred
Gemcitabine monophosphate is an active intermediate of Gemcitabine, designed for research use. It exhibits a synergistic anti-cancer effect and can be delivered by formulating it into nanoparticles. It has been shown to induce tumor cell apoptosis and reduce tumor cell proliferation.
  • Has a synergistic anti-cancer effect
  • Can be delivered by formulating it into nanoparticles
  • Induces tumor cell apoptosis (30-40%)
  • Reduces tumor cell proliferation (by 8 times)
  • Significantly reduces tumor microvessel density (MVD)
  • Inhibits tumor mitosis
Catalog No. 50-003-45774 Supplier Medchemexpress LLC Supplier No. HY108932100MG
Gemcitabine monophosphate is an active intermediate of Gemcitabine, designed for research use. It exhibits a synergistic anti-cancer effect and can be delivered by formulating it into nanoparticles. It has been shown to induce tumor cell apoptosis and reduce tumor cell proliferation.
  • Has a synergistic anti-cancer effect
  • Can be delivered by formulating it into nanoparticles
  • Induces tumor cell apoptosis (30-40%)
  • Reduces tumor cell proliferation (by 8 times)
  • Significantly reduces tumor microvessel density (MVD)
  • Inhibits tumor mitosis