Learn More
Medchemexpress LLC Gemcitabine monophosphate disodium | 1638288-31-9 | 387.14 | 10 MM * 1 ML
SDP
Catalog No. 5000345973
Encompass_Preferred
Gemcitabine monophosphate (Gemcitabine 5′-phosphate) is one of the active intermediates of Gemcitabine. It has a synergistic anti-cancer effect and can be delivered by formulating it into nanoparticles.
  • Active intermediate of Gemcitabine.
  • Synergistic anti-cancer effect.
  • Can be delivered via nanoparticles.
  • In vivo studies show it induces tumor cell apoptosis (30-40%), reduces tumor cell proliferation by 8 times, significantly reduces tumor microvessel density (MVD), and inhibits tumor mitosis in subcutaneous and orthotopic xenograft mouse models of non-small cell lung cancer.
Catalog No. 50-003-45973 Supplier Medchemexpress LLC Supplier No. HY10893250MG
Gemcitabine monophosphate (Gemcitabine 5′-phosphate) is one of the active intermediates of Gemcitabine. It has a synergistic anti-cancer effect and can be delivered by formulating it into nanoparticles.
  • Active intermediate of Gemcitabine.
  • Synergistic anti-cancer effect.
  • Can be delivered via nanoparticles.
  • In vivo studies show it induces tumor cell apoptosis (30-40%), reduces tumor cell proliferation by 8 times, significantly reduces tumor microvessel density (MVD), and inhibits tumor mitosis in subcutaneous and orthotopic xenograft mouse models of non-small cell lung cancer.