Learn More
Medchemexpress LLC Gemcitabine-o-si(di-iso)-o-mc | 98.8% | 558.65 g·mol⁻1 | C24H36F2N4O7Si | 5 MG
SDP
Catalog No. 5000203239
Encompass_Preferred
Gemcitabine-O-Si(di-iso)-O-Mc is a gemcitabine-based drug-linker conjugate supplied for antibody-drug conjugate research. It couples the cytotoxic nucleoside analog gemcitabine to an Si(di-iso)-based ADC linker and is provided as a characterized, research-grade reagent.
  • Intended for antibody-drug conjugate research and development.
  • Reported purity 98.8%.
  • Molecular weight 558.65 g·mol⁻1 and formula C24H36F2N4O7Si.
  • Available in small-mass quantities suitable for lab-scale use.
  • Provided with batch-specific certificate of analysis.
Catalog No. 50-002-03239 Supplier Medchemexpress LLC Supplier No. HY1308125MG
Gemcitabine-O-Si(di-iso)-O-Mc is a gemcitabine-based drug-linker conjugate supplied for antibody-drug conjugate research. It couples the cytotoxic nucleoside analog gemcitabine to an Si(di-iso)-based ADC linker and is provided as a characterized, research-grade reagent.
  • Intended for antibody-drug conjugate research and development.
  • Reported purity 98.8%.
  • Molecular weight 558.65 g·mol⁻1 and formula C24H36F2N4O7Si.
  • Available in small-mass quantities suitable for lab-scale use.
  • Provided with batch-specific certificate of analysis.