Learn More
Medchemexpress LLC Gemcitabine-o-si(di-iso)-o-mc | 98.8% | 558.65 g/mol | C24H36F2N4O7Si | 1 MG
SDP
Catalog No. 5000203233
Encompass_Preferred
Gemcitabine-O-Si(di-iso)-O-Mc is a drug-linker conjugate based on gemcitabine intended for antibody-drug conjugate research and bioconjugation workflows. It is supplied as a solid in small research quantities and is used for medicinal chemistry, ADC synthesis, and preclinical evaluation.
  • Drug-linker conjugate for ADC research.
  • Supplied as a solid suitable for conjugation and analytical characterization.
  • High purity (98.8%) for research applications.
  • Store at -20°C and protect from light; follow handling instructions for solvent storage.
  • Available in small quantities (e.g., 1 mg) for method development and synthesis work.
Catalog No. 50-002-03233 Supplier Medchemexpress LLC Supplier No. HY1308121MG
Gemcitabine-O-Si(di-iso)-O-Mc is a drug-linker conjugate based on gemcitabine intended for antibody-drug conjugate research and bioconjugation workflows. It is supplied as a solid in small research quantities and is used for medicinal chemistry, ADC synthesis, and preclinical evaluation.
  • Drug-linker conjugate for ADC research.
  • Supplied as a solid suitable for conjugation and analytical characterization.
  • High purity (98.8%) for research applications.
  • Store at -20°C and protect from light; follow handling instructions for solvent storage.
  • Available in small quantities (e.g., 1 mg) for method development and synthesis work.