Learn More
Perkin Elmer US LLC Sample Capillary Adapter for GemCone and GemTip Nebulizers
Catalog No. NC1875955
Encompass_Preferred
This Capillary Adapter, or Gland Nut, is included in the GemTip High-Sensitivity Plastic Body Nebulizer Complete Assembly for AAnalyst (N0370393) and the Universal GemTip Corrosion-Resistant Plastic Body Nebulizer Complete Assembly for AAnalyst (N0370394). It is used in the GemCone Low Flow Nebulizer (N0770358) and the GemCone High Dissolved Solids Nebulizer (N0690670).
This item is not returnable. View return policy
Catalog No. NC1875955 Supplier Perkin Elmer US LLC Supplier No. N0371505
This Capillary Adapter, or Gland Nut, is included in the GemTip High-Sensitivity Plastic Body Nebulizer Complete Assembly for AAnalyst (N0370393) and the Universal GemTip Corrosion-Resistant Plastic Body Nebulizer Complete Assembly for AAnalyst (N0370394). It is used in the GemCone Low Flow Nebulizer (N0770358) and the GemCone High Dissolved Solids Nebulizer (N0690670).