Learn More
Perkin Elmer US LLC GemCone Low-Flow Nebulizer for Optima 2x00/4x00/5x00/7x00/8x00 and Avio 200/500
Catalog No. 509043783
Encompass_Preferred
The nebulizer is optimized for low nebulizer gas flow for use with cyclonic spray chambers. It requires an end cap (N0680504) for use with Ryton™ Scott spray chambers. This GemCone Nebulizer is included in the Organics Sample Introduction System for Avio 200 (N0790611), Organics Sample Introduction System for Avio 500 (N0810609), Quick-Change Torch Module for Oil Analysis for Optima 4300V/5300V/7300V (N0770910), Organics Kit for Avio 200/500 (N0790610), Organics Kit for Optima 8x00 (N0780608), Organics Kit for Optima 2x00/4x00/5x00/7x00 DV (N0770420), and the Organics Kit for Optima 4300 V/5300 V/7300 V (N0770732).
Catalog No. 50-904-3783 Supplier Perkin Elmer US LLC Supplier No. N0770358
The nebulizer is optimized for low nebulizer gas flow for use with cyclonic spray chambers. It requires an end cap (N0680504) for use with Ryton™ Scott spray chambers. This GemCone Nebulizer is included in the Organics Sample Introduction System for Avio 200 (N0790611), Organics Sample Introduction System for Avio 500 (N0810609), Quick-Change Torch Module for Oil Analysis for Optima 4300V/5300V/7300V (N0770910), Organics Kit for Avio 200/500 (N0790610), Organics Kit for Optima 8x00 (N0780608), Organics Kit for Optima 2x00/4x00/5x00/7x00 DV (N0770420), and the Organics Kit for Optima 4300 V/5300 V/7300 V (N0770732).