Learn More
Perkin Elmer US LLC GemCone High Dissolved Solids Nebulizer
Catalog No. 509043700
Encompass_Preferred
GemCone Nebulizers require an end cap (N0680343) for use with Scott Spray Chambers. This GemCone Nebulizer is included in the Quick-Change Torch Module for HF Analysis for Optima 4300V/5300V/7300V (N0770911).
Catalog No. 50-904-3700 Supplier Perkin Elmer US LLC Supplier No. N0690670
GemCone Nebulizers require an end cap (N0680343) for use with Scott Spray Chambers. This GemCone Nebulizer is included in the Quick-Change Torch Module for HF Analysis for Optima 4300V/5300V/7300V (N0770911).