Learn More
Medchemexpress LLC HY-129993 5mg , Gemfibrozil 1-O-β-glucuronide CAS:91683-38-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1299935MG
Encompass_Preferred
HY-129993 5mg Gemfibrozil 1-O-β-glucuronide CAS: 91683-38-4 Gemfibrozil 1-O-β-Glucuronide, a metabolite of Gemfibrozil (CI-719; HY-B0258), is a potent and competitive P450 (CYP) isoform CYP2C8 inhibitor with an IC50 of 4.07 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1747