Learn More
Medchemexpress LLC HY-B0258 100mg Medchemexpress, Gemfibrozil CAS:25812-30-0 Purity:>98%
SDP
Catalog No. 502082428
Encompass_Preferred
Medchemexpress, HY-B0258 100mg Gemfibrozil CAS:25812-30-0 Gemfibrozil is an activator of PPAR-α, used as a lipid-lowering drug; Gemfibrozil is also a nonselective inhibitor of several P450 isoforms, with Ki values for CYP2C9, 2C19, 2C8, and 1A2 of 5.8, 24, 69, and 82 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2428 Supplier Medchemexpress LLC Supplier No. HYB0258100MG
Medchemexpress, HY-B0258 100mg Gemfibrozil CAS:25812-30-0 Gemfibrozil is an activator of PPAR-α, used as a lipid-lowering drug; Gemfibrozil is also a nonselective inhibitor of several P450 isoforms, with Ki values for CYP2C9, 2C19, 2C8, and 1A2 of 5.8, 24, 69, and 82 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.