Learn More
Apexbio Technology LLC Gemfibrozil 25812-30-0 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B19475.1
Encompass_Preferred
Gemfibrozil is a small-molecule pharmacological agent that functions primarily through activation of the nuclear receptor peroxisome proliferator-activated receptor-alpha (PPAR ) By modulating PPAR signaling pathways gemfibrozil regulates lipid metabolism influencing the transcription of genes associated with fatty acid oxidation and lipoprotein synthesis resulting in reduced triglyceride levels and altered lipoprotein profiles In biomedical research contexts gemfibrozil is utilized to explore molecular mechanisms underlying lipid metabolic processes atherosclerosis progression and related metabolic disorders Furthermore gemfibrozil serves as a useful experimental tool to dissect PPAR -dependent pathways and to investigate therapeutic potentials targeting lipid abnormalities and cardiovascular disease
Catalog No. 50-000-62354