Learn More
Medchemexpress LLC Gemifloxacin mesylate | 210353-53-0 | 99.59% | 5 MG
SDP
Catalog No. 5000411742
Encompass_Preferred
Gemifloxacin mesylate (SB-265805S; LB-20304a) is an orally active broad-spectrum quinolone antibacterial antibiotic. It inhibits DNA synthesis by inhibiting DNA gyrase and Topoisomerase IV activities. Gemifloxacin mesylate has potent antibacterial activities against gram-positive bacteria in in vitro efficacy study, particularly Streptococci and Staphylococci. It has been used in the research of respiratory tract infections.
  • Inhibits DNA synthesis by inhibiting DNA gyrase and Topoisomerase IV activities.
  • Potent antibacterial activities against gram-positive bacteria in in vitro efficacy study, particularly Streptococci and Staphylococci.
  • Used in the research of respiratory tract infections.
Catalog No. 50-004-11742 Supplier Medchemexpress LLC Supplier No. HYB10505MG
Gemifloxacin mesylate (SB-265805S; LB-20304a) is an orally active broad-spectrum quinolone antibacterial antibiotic. It inhibits DNA synthesis by inhibiting DNA gyrase and Topoisomerase IV activities. Gemifloxacin mesylate has potent antibacterial activities against gram-positive bacteria in in vitro efficacy study, particularly Streptococci and Staphylococci. It has been used in the research of respiratory tract infections.
  • Inhibits DNA synthesis by inhibiting DNA gyrase and Topoisomerase IV activities.
  • Potent antibacterial activities against gram-positive bacteria in in vitro efficacy study, particularly Streptococci and Staphylococci.
  • Used in the research of respiratory tract infections.