Learn More
Medchemexpress LLC Gemifloxacin (mesylate) | 210353-53-0 | 99.7% | 50 MG
SDP
Catalog No. 5000413861
Encompass_Preferred
Gemifloxacin mesylate is an orally active, broad-spectrum quinolone antibacterial antibiotic that inhibits DNA synthesis by targeting DNA gyrase and Topoisomerase IV activities. It exhibits potent antibacterial activities against gram-positive bacteria, particularly Streptococci and Staphylococci, and has been utilized in research for respiratory tract infections.
  • Inhibits DNA gyrase and Topoisomerase IV activities
  • Potent antibacterial activities against gram-positive bacteria
  • Effective against Streptococci and Staphylococci
  • Used in respiratory tract infection research
Catalog No. 50-004-13861 Supplier Medchemexpress LLC Supplier No. HYB105050MG
Gemifloxacin mesylate is an orally active, broad-spectrum quinolone antibacterial antibiotic that inhibits DNA synthesis by targeting DNA gyrase and Topoisomerase IV activities. It exhibits potent antibacterial activities against gram-positive bacteria, particularly Streptococci and Staphylococci, and has been utilized in research for respiratory tract infections.
  • Inhibits DNA gyrase and Topoisomerase IV activities
  • Potent antibacterial activities against gram-positive bacteria
  • Effective against Streptococci and Staphylococci
  • Used in respiratory tract infection research