Learn More
Medchemexpress LLC Gemifloxacin mesylate | 210353-53-0 | MFCD08277637 | 99.59% | 1 ML
SDP
Catalog No. 5000413748
Encompass_Preferred
Gemifloxacin mesylate is an orally active, broad-spectrum quinolone antibacterial antibiotic. It works by inhibiting DNA synthesis through the inhibition of DNA gyrase and Topoisomerase IV activities. It has been utilized in the research of respiratory tract infections.
  • Inhibits DNA synthesis by targeting DNA gyrase and Topoisomerase IV activities
  • Exhibits potent antibacterial activities against gram-positive bacteria in vitro, particularly Streptococci and Staphylococci
  • Higher antibacterial activity than Moxifloxacin against Streptococcus pneumoniae with a MIC90 of 0.06 μg/mL
  • Highly potent antibacterial activity against Penicillin-resistant strains of S. pneumoniae with a MIC90 of 0.03 μg/mL
Catalog No. 50-004-13748 Supplier Medchemexpress LLC Supplier No. HYB105010MM/1ML
Gemifloxacin mesylate is an orally active, broad-spectrum quinolone antibacterial antibiotic. It works by inhibiting DNA synthesis through the inhibition of DNA gyrase and Topoisomerase IV activities. It has been utilized in the research of respiratory tract infections.
  • Inhibits DNA synthesis by targeting DNA gyrase and Topoisomerase IV activities
  • Exhibits potent antibacterial activities against gram-positive bacteria in vitro, particularly Streptococci and Staphylococci
  • Higher antibacterial activity than Moxifloxacin against Streptococcus pneumoniae with a MIC90 of 0.06 μg/mL
  • Highly potent antibacterial activity against Penicillin-resistant strains of S. pneumoniae with a MIC90 of 0.03 μg/mL