Learn More
TARGETMOL CHEMICALS INC Gemifloxacin mesylate 50MG
SDP
Catalog No. 502878537
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Gemifloxacin mesylate (Gemifloxacinmesylate) inhibits activities of DNA gyrase and topoisomerase IV, thereby inhibiting DNA replication and eventually bacterial growth. Gemifloxacin Mesylate is the mesylate salt form of gemifloxacin, a synthetic broad-spectrum fluoroquinolone with antibacterial activity. This fluoroquinolone exerts an enhanced spectrum of activity against gram-positive bacteria such as Streptococcus pneumonia and Staphylococcus aureus, in addition to its activity against gram-negative bacteria. Purity 99.55%
Catalog No. 50-287-8537 Supplier TARGETMOL CHEMICALS INC Supplier No. T149250MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Gemifloxacin mesylate (Gemifloxacinmesylate) inhibits activities of DNA gyrase and topoisomerase IV, thereby inhibiting DNA replication and eventually bacterial growth. Gemifloxacin Mesylate is the mesylate salt form of gemifloxacin, a synthetic broad-spectrum fluoroquinolone with antibacterial activity. This fluoroquinolone exerts an enhanced spectrum of activity against gram-positive bacteria such as Streptococcus pneumonia and Staphylococcus aureus, in addition to its activity against gram-negative bacteria. Purity 99.55%