Learn More
Selleck Chemical LLC Gemifloxacin mesylate
SDP
Catalog No. 503136430
Encompass_Preferred
Gemifloxacin mesylate (SB265805 mesylate LB20304 mesylate) is a novel fluoronaphthyridone with a broad spectrum of antimicrobial activity and enhances activity against gram-positive aerobes
Catalog No. 50-313-6430 Supplier Selleck Chemical LLC Supplier No. E07065MG
Gemifloxacin mesylate (SB265805 mesylate LB20304 mesylate) is a novel fluoronaphthyridone with a broad spectrum of antimicrobial activity and enhances activity against gram-positive aerobes