Learn More
Cayman Chemical Gemifloxacin (mesylate), Purity: ≥98%, CAS: 210353-53-0, 50 mg
SDP
Catalog No. 502984073
Encompass_Preferred
A fluoroquinolone antibiotic that is effective against C. pneumoniae and M. tuberculosis (MIC50s = 0.25 and 8 µg/ml, respectively); useful against respiratory tract infections
Catalog No. 50-298-4073 Supplier Cayman Chemical Supplier No. 2104750 MG
A fluoroquinolone antibiotic that is effective against C. pneumoniae and M. tuberculosis (MIC50s = 0.25 and 8 µg/ml, respectively); useful against respiratory tract infections