Learn More
Medchemexpress LLC Gemigliptin (tartrate) | 1374639-74-3 | 99.5% | 639.45 | 1 ML
SDP
Catalog No. 5000359503
Encompass_Preferred
Gemigliptin tartrate, also known as LC15-0444 tartrate, is a highly selective, reversible, and competitive dipeptidyl peptidase-4 (DPP-4) inhibitor with an IC50 of 10.3 nM for human recombinant DPP-4. It demonstrates potent anti-glycation properties and can be used in research concerning advanced glycation end products (AGE)-related diabetic complications.
  • Highly selective DPP-4 inhibitor.
  • Exhibits potent anti-glycation properties.
  • Useful for research on AGE-related diabetic complications.
  • Dose-dependently inhibits AGE-BSA formation.
  • Suppresses cross-linking of preformed AGE-BSA.
Catalog No. 50-003-59503 Supplier Medchemexpress LLC Supplier No. HY14892A10MM/1ML
Gemigliptin tartrate, also known as LC15-0444 tartrate, is a highly selective, reversible, and competitive dipeptidyl peptidase-4 (DPP-4) inhibitor with an IC50 of 10.3 nM for human recombinant DPP-4. It demonstrates potent anti-glycation properties and can be used in research concerning advanced glycation end products (AGE)-related diabetic complications.
  • Highly selective DPP-4 inhibitor.
  • Exhibits potent anti-glycation properties.
  • Useful for research on AGE-related diabetic complications.
  • Dose-dependently inhibits AGE-BSA formation.
  • Suppresses cross-linking of preformed AGE-BSA.