Learn More
Medchemexpress LLC Gemigliptin tartrate | 1374639-74-3 | 99.5% | 639.45 | 100 MG
SDP
Catalog No. 5000359451
Encompass_Preferred
Gemigliptin tartrate is a highly selective, reversible, and competitive dipeptidyl peptidase-4 (DPP-4) inhibitor. It possesses potent anti-glycation properties and is suitable for research into advanced glycation end products (AGE)-related diabetic complications.
  • Highly selective, reversible, and competitive DPP-4 inhibitor
  • IC50 of 10.3 nM for human recombinant DPP-4
  • Exhibits potent anti-glycation properties
  • Inhibits formation of AGE-BSA
  • Suppresses cross-linking of preformed AGE-BSA
  • Inhibits AGEs formation and AGE cross-links in vivo
Catalog No. 50-003-59451 Supplier Medchemexpress LLC Supplier No. HY14892A100MG
Gemigliptin tartrate is a highly selective, reversible, and competitive dipeptidyl peptidase-4 (DPP-4) inhibitor. It possesses potent anti-glycation properties and is suitable for research into advanced glycation end products (AGE)-related diabetic complications.
  • Highly selective, reversible, and competitive DPP-4 inhibitor
  • IC50 of 10.3 nM for human recombinant DPP-4
  • Exhibits potent anti-glycation properties
  • Inhibits formation of AGE-BSA
  • Suppresses cross-linking of preformed AGE-BSA
  • Inhibits AGEs formation and AGE cross-links in vivo