Learn More
Medchemexpress LLC Gemigliptin tartrate | 1374639-74-3 | 99.5% | 639.45 | 5 MG
SDP
Catalog No. 5000361142
Encompass_Preferred
Gemigliptin tartrate is a highly selective, reversible, and competitive dipeptidyl peptidase-4 (DPP-4) inhibitor, with an IC50 of 10.3 nM for human recombinant DPP-4. It exhibits potent anti-glycation properties and can be used for the research of advanced glycation end products (AGE)-related diabetic complications. This product is for research use only.
  • Highly selective, reversible, and competitive dipeptidyl peptidase-4 (DPP-4) inhibitor
  • IC50 of 10.3 nM for human recombinant DPP-4
  • Exhibits potent anti-glycation properties
  • Useful for research of advanced glycation end products (AGE)-related diabetic complications
  • Dose-dependently inhibits the formation of AGE-BSA
  • Suppresses the cross-linking of preformed AGE-BSA with rat tail tendon collagen
  • Inhibits AGEs formation and AGE cross-links in vivo
  • Dose-dependently inhibits plasma DPP-4 activity in various animal models
Catalog No. 50-003-61142 Supplier Medchemexpress LLC Supplier No. HY14892A5MG
Gemigliptin tartrate is a highly selective, reversible, and competitive dipeptidyl peptidase-4 (DPP-4) inhibitor, with an IC50 of 10.3 nM for human recombinant DPP-4. It exhibits potent anti-glycation properties and can be used for the research of advanced glycation end products (AGE)-related diabetic complications. This product is for research use only.
  • Highly selective, reversible, and competitive dipeptidyl peptidase-4 (DPP-4) inhibitor
  • IC50 of 10.3 nM for human recombinant DPP-4
  • Exhibits potent anti-glycation properties
  • Useful for research of advanced glycation end products (AGE)-related diabetic complications
  • Dose-dependently inhibits the formation of AGE-BSA
  • Suppresses the cross-linking of preformed AGE-BSA with rat tail tendon collagen
  • Inhibits AGEs formation and AGE cross-links in vivo
  • Dose-dependently inhibits plasma DPP-4 activity in various animal models