Learn More
Medchemexpress LLC Gemigliptin tartrate | 1374639-74-3 | 99.5% | 639.45 | 50 MG
SDP
Catalog No. 5000359569
Encompass_Preferred
Gemigliptin tartrate is a highly selective, reversible, and competitive dipeptidyl peptidase-4 (DPP-4) inhibitor with an IC50 of 10.3 nM for human recombinant DPP-4. It exhibits potent anti-glycation properties and can be utilized for research into advanced glycation end products (AGE)-related diabetic complications.
  • Highly selective, reversible, and competitive DPP-4 inhibitor
  • Exhibits potent anti-glycation properties
  • Can be used for research into advanced glycation end products (AGE)-related diabetic complications
  • Inhibits the formation of AGE-BSA
  • Suppresses the cross-linking of preformed AGE-BSA
  • Dose-dependently inhibits plasma DPP-4 activity in rats, dogs, and monkeys
  • Significantly reduces circulating AGE levels in serum in vivo
Catalog No. 50-003-59569 Supplier Medchemexpress LLC Supplier No. HY14892A50MG
Gemigliptin tartrate is a highly selective, reversible, and competitive dipeptidyl peptidase-4 (DPP-4) inhibitor with an IC50 of 10.3 nM for human recombinant DPP-4. It exhibits potent anti-glycation properties and can be utilized for research into advanced glycation end products (AGE)-related diabetic complications.
  • Highly selective, reversible, and competitive DPP-4 inhibitor
  • Exhibits potent anti-glycation properties
  • Can be used for research into advanced glycation end products (AGE)-related diabetic complications
  • Inhibits the formation of AGE-BSA
  • Suppresses the cross-linking of preformed AGE-BSA
  • Dose-dependently inhibits plasma DPP-4 activity in rats, dogs, and monkeys
  • Significantly reduces circulating AGE levels in serum in vivo