Learn More
Medchemexpress LLC Gemigliptin tartrate hydrate | 1375415-82-9 | 99.8% | 666.49 | 100 MG
SDP
Catalog No. 5000359391
Encompass_Preferred
Gemigliptin (LC15-0444) tartrate hydrate is a selective, reversible, and competitive dipeptidyl peptidase-4 (DPP-4) inhibitor, with an IC50 of 10.3 nM for human recombinant DPP-4. It exhibits potent anti-glycation properties and can be used for the research of advanced glycation end products (AGE)-related diabetic complications.
  • Selective, reversible, and competitive dipeptidyl peptidase-4 (DPP-4) inhibitor
  • Exhibits potent anti-glycation properties
  • Used for the research of advanced glycation end products (AGE)-related diabetic complications
Catalog No. 50-003-59391 Supplier Medchemexpress LLC Supplier No. HY14892B100MG
Gemigliptin (LC15-0444) tartrate hydrate is a selective, reversible, and competitive dipeptidyl peptidase-4 (DPP-4) inhibitor, with an IC50 of 10.3 nM for human recombinant DPP-4. It exhibits potent anti-glycation properties and can be used for the research of advanced glycation end products (AGE)-related diabetic complications.
  • Selective, reversible, and competitive dipeptidyl peptidase-4 (DPP-4) inhibitor
  • Exhibits potent anti-glycation properties
  • Used for the research of advanced glycation end products (AGE)-related diabetic complications