Learn More
Medchemexpress LLC Gemigliptin tartrate hydrate | 1375415-82-9 | 99.8% | 666.49 | 25 MG
SDP
Catalog No. 5000363475
Encompass_Preferred
Gemigliptin tartrate hydrate is a selective, reversible, and competitive dipeptidyl peptidase-4 (DPP-4) inhibitor. It exhibits potent anti-glycation properties and can be used for the research of advanced glycation end products (AGE)-related diabetic complications.
  • Selective, reversible, and competitive dipeptidyl peptidase-4 (DPP-4) inhibitor
  • Exhibits potent anti-glycation properties
  • Used for research of advanced glycation end products (AGE)-related diabetic complications
  • IC50 of 10.3 nM for human recombinant DPP-4
Catalog No. 50-003-63475 Supplier Medchemexpress LLC Supplier No. 25MGHY14892B
Gemigliptin tartrate hydrate is a selective, reversible, and competitive dipeptidyl peptidase-4 (DPP-4) inhibitor. It exhibits potent anti-glycation properties and can be used for the research of advanced glycation end products (AGE)-related diabetic complications.
  • Selective, reversible, and competitive dipeptidyl peptidase-4 (DPP-4) inhibitor
  • Exhibits potent anti-glycation properties
  • Used for research of advanced glycation end products (AGE)-related diabetic complications
  • IC50 of 10.3 nM for human recombinant DPP-4