Learn More
Medchemexpress LLC Gemigliptin tartrate hydrate | 1375415-82-9 | 99.8% | 666.49 | C18H19F8N5O2·C4H6O6·3/2H2O | 10 MG
SDP
Catalog No. 5000285368
Encompass_Preferred
Gemigliptin tartrate hydrate is a selective, reversible, competitive dipeptidyl peptidase-4 (DPP-4) inhibitor supplied as a white to off-white solid hydrate for research use. It is provided in research-scale packages with high chemical purity and characterized molecular weight, appropriate for biochemical and cellular studies, including investigations of advanced glycation end products and diabetic complications.
  • Selective inhibitor of DPP-4 with low nanomolar potency.
  • High purity suitable for analytical and biological research.
  • Solid white to off-white appearance for easy handling.
  • Hydrated salt form with defined molecular weight for reproducibility.
  • Recommended storage conditions help preserve stability.
  • Available in small, research-scale package sizes for laboratory use.
Catalog No. 50-002-85368 Supplier Medchemexpress LLC Supplier No. HY14892B10MG
Gemigliptin tartrate hydrate is a selective, reversible, competitive dipeptidyl peptidase-4 (DPP-4) inhibitor supplied as a white to off-white solid hydrate for research use. It is provided in research-scale packages with high chemical purity and characterized molecular weight, appropriate for biochemical and cellular studies, including investigations of advanced glycation end products and diabetic complications.
  • Selective inhibitor of DPP-4 with low nanomolar potency.
  • High purity suitable for analytical and biological research.
  • Solid white to off-white appearance for easy handling.
  • Hydrated salt form with defined molecular weight for reproducibility.
  • Recommended storage conditions help preserve stability.
  • Available in small, research-scale package sizes for laboratory use.