Learn More
Cayman Chemical GemIglIptIntartrate 10mg
SDP
Catalog No. 502911098
Encompass_Preferred
A DPP-4 inhibitor (IC50 = 0.01 µM); selective for DPP-4 over DPP-8, DPP-9, and FAP- α (IC50s = 277.28, 233.72, 418.43 µM, respectively); inhibits DPP-4 activity in isolated human plasma (IC50 = 11.5 nM); scavenges methylglyoxal in vitro and inhibits AGE formation between BSA and methylglyoxal and cross-linking of AGE-BSA with rat tail tendon collagen (IC50s = 1.15, 11.69, and 1.39 mM, respectively); reduces the levels of circulating AGE and red blood cell IgG content in db/db diabetic mice at 100 mg/kg; decreases blood glucose and HbA1c levels and reduces pancreatic β-cell damage in a mouse model of STZ- and high-fat diet-induced diabetes; induces cytotoxicity in SW1736 and TPC-1 cells
Catalog No. 50-291-1098 Supplier Cayman Chemical Supplier No. 3601610 MG
A DPP-4 inhibitor (IC50 = 0.01 µM); selective for DPP-4 over DPP-8, DPP-9, and FAP- α (IC50s = 277.28, 233.72, 418.43 µM, respectively); inhibits DPP-4 activity in isolated human plasma (IC50 = 11.5 nM); scavenges methylglyoxal in vitro and inhibits AGE formation between BSA and methylglyoxal and cross-linking of AGE-BSA with rat tail tendon collagen (IC50s = 1.15, 11.69, and 1.39 mM, respectively); reduces the levels of circulating AGE and red blood cell IgG content in db/db diabetic mice at 100 mg/kg; decreases blood glucose and HbA1c levels and reduces pancreatic β-cell damage in a mouse model of STZ- and high-fat diet-induced diabetes; induces cytotoxicity in SW1736 and TPC-1 cells