Learn More
Medchemexpress LLC HY-16780 5mg Medchemexpress, Gemilukast CAS:1232861-58-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY167805MG
Encompass_Preferred
Medchemexpress, HY-16780 5mg Gemilukast CAS:1232861-58-3 Gemilukast is an orally active and potent dual cysteinyl leukotriene 1 and 2 receptors (CysLT 1 and CysLT 2 ) antagonist, with IC 50 s of 1.7, 25 nM for human CysLT 1 and CysLT 2 , respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2782