Learn More
Aviva Systems Biology Corp GEMIN6 Antibody - middle region (ARP62117_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62117_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GEMIN6. It was validated on Western Blot. . Alternate names: FLJ23459ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-287-576