Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc LC Column 75 x 21.2 mm

Supplier:  Phenomenex Inc 00C4763P0AX

Encompass

Gemini 10u C8(3) 100A, AXIA Packed 00C-4763-P0-AX

Catalog No. 50-239-3572


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.