Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc LC Column 125 x 4.6 mm

Catalog No. 502393575 Shop All Phenomenex Products
Encompass

Gemini 5 m NX-C18 110 00E-4454-E0

Catalog No. 50-239-3575 Supplier Phenomenex Inc Supplier No. 00E4454E0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gemini 5 m NX-C18 110 00E-4454-E0

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.