Learn More
Stoelting™ Gemini Active and Passive Avoidance System
SDP
Catalog No. p-7163558 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse
Mouse, Rat
Rat
Model:
Shock
Shock and Additional stations
Shock and Air stimulation
Shock, Air stim., Add'l Stations
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Animal Model
10-002-008 Mouse Shock
10-002-009 Rat Shock
10-001-003 Mouse, Rat Shock
10-001-004 Mouse, Rat Shock and Additional stations
10-002-010 Mouse Shock and Air stimulation
10-002-011 Rat Shock and Air stimulation
10-001-005 Mouse, Rat Shock and Air stimulation
10-002-012 Mouse Shock, Air stim., Add'l Stations
10-002-013 Rat Shock, Air stim., Add'l Stations
10-001-006 Mouse, Rat Shock, Air stim., Add'l Stations
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 10-002-008 Supplier Stoelting™ Supplier No. 57875M
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Gemini Active and Passive Avoidance System was designed specifically to test active and passive avoidance in rats and mice.
  • Unique design allows users to run up to 8 independently operated stations directly from a laptop using a standard USB.
  • By allowing users to define a complete study in advance, Gemini eliminates run time data entry that could affect the accuracy of results.
  • Scrambled shock provides more accurate responses
    • Solid state feedback-controlled, constant current shocker uses 8 unique outputs to scramble the shock across the grid floor.
    • Scrambling the shock prevents the animal from determining a pattern and leads to more accurate responses.
  • Kinder, gentler, unconditioned stimulus - offers an alternative air puff unconditioned stimulus
  • Optional Components
    • Air puff stimulation unit (AIRSTIM)
    • Additional test stations
    • Mouse passive avoidance start box
    • Grid floors (for rats or mice)
Specifications
For Use With (Application) Neuroscience
Animal Mouse
Model Shock