Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Gemini™ Chemical Identification Analyzer
GSA_VA

Catalog No. 18001145 Shop All Thermo Scientific Products
Change view
Click to view available options
Includes:
1 Year of Support
2 Years of Support
3 Years of Support
4 Years of Support
5 years of support
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
18-001-145 1 Year of Support
18-001-146 2 Years of Support
18-001-147 3 Years of Support
18-001-148 4 Years of Support
18-001-149 5 years of support
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-001-145 Supplier Thermo Scientific™ Supplier No. 8000501101

To request a quote for this item, please select Request a Quote to reach our dedicated support team.

missing translation for 'validMainframeMsg'

Trusted by agencies worldwide since its launch in 2015, the Thermo Scientific™ Gemini™ Analyzer identifies unknown chemicals in the field using Raman and FTIR in a single, handheld device.

Since its launch in 2015, the Thermo Scientific™ Gemini™ Chemical Identification Analyzer has been trusted by elite military units, public safety forces, and first responders around the world. Built on the performance of the Thermo Scientific™ FirstDefender™ and TruDefender™ Analyzers, the Gemini™ analyzer is designed for reliable performance in harsh environments. Its rugged, compact design is engineered to meet mission-critical demands, helping operators make faster, better-informed decisions in the field.

Accuracy and reliability with advanced algorithms

The Gemini™ analyzer combines both Raman and FTIR technologies in a single handheld instrument, enabling complementary testing on scene. Leveraging advanced algorithms and a comprehensive onboard library, it supports the accurate identification of unknown solids and liquids, including explosives, chemical warfare agents, and industrial chemicals. Features like onboard mixture analysis and 24/7 reachback support reinforce operator confidence.

Ease of use for field operations

Designed for operators who work under pressure, Scan Assist technology guides users through sampling and technology selection, while customizable profiles assist the user with minimizing user error. Features like the motorized anvil and Scan Delay enhance safety by allowing operators to maintain distance from potential hazards. With intuitive workflows and a compact form factor, the Gemini™ analyzer reduces the burden of heavy equipment while increasing operational efficiency.

Add-on

Hazmaster G3: Integrated HazMasterG3® decision support software helps operators interpret results and assess potential threats from identified chemicals, supporting actionable insights directly in the field.

LowDoseID: By utilizing the enhanced hardware and software of the Gemini™ analyzer, combined with SERS, the Gemini™ analyzer with LowDoseID enables low concentration analysis and results for key narcotics.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certified to MIL-STD-810G and IP67; full test report available on request
Collection Optics FTIR: ATR diamond crystal, single bounce, Raman: NA = 0.33
Data Export Formats SPC file (for use in standard spectroscopic software), text file, or JPEG report
Dimensions (L x W x H) 10.1 x 5.7 x 2.4 in. (25.6 x 14.6 x 6.1 cm)
Includes 1 Year of Support
IP Rating IP67
Languages English
Power Supply Internal 3.7 V Lithium Polymer Battery Pack ( QTY 1), Internal CR123A Batteries (QTY 3), DC Wall Adapter, 12 V DC, 1.25 A
Spectral Range FTIR: 4,000 cm-1 to 650 cm-1, Raman: 250 cm-1 to 2875 cm-1
Temperature Range (Metric) -20°C to +50°C
Weight (English) 4.2 lb.
Weight (Metric) 1.9 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.