Learn More
Thermo Scientific™ Gemini™ Chemical Identification Analyzer
GSA_VA
Catalog No. 18001149 Shop All Thermo Scientific Products
Change view
Click to view available options
Includes:
1 Year of Support
2 Years of Support
3 Years of Support
4 Years of Support
5 years of support
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
18-001-149 5 years of support
18-001-145 1 Year of Support
18-001-146 2 Years of Support
18-001-147 3 Years of Support
18-001-148 4 Years of Support
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 18-001-149 Supplier Thermo Scientific™ Supplier No. 8000501501
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
This item is not returnable. View return policy
Trusted by agencies worldwide since its launch in 2015, the Thermo Scientific™ Gemini™ Analyzer identifies unknown chemicals in the field using Raman and FTIR in a single, handheld device.
Since its launch in 2015, the Thermo Scientific™ Gemini™ Chemical Identification Analyzer has been trusted by elite military units, public safety forces, and first responders around the world. Built on the performance of the Thermo Scientific™ FirstDefender™ and TruDefender™ Analyzers, the Gemini™ analyzer is designed for reliable performance in harsh environments. Its rugged, compact design is engineered to meet mission-critical demands, helping operators make faster, better-informed decisions in the field.
Accuracy and reliability with advanced algorithms
The Gemini™ analyzer combines both Raman and FTIR technologies in a single handheld instrument, enabling complementary testing on scene. Leveraging advanced algorithms and a comprehensive onboard library, it supports the accurate identification of unknown solids and liquids, including explosives, chemical warfare agents, and industrial chemicals. Features like onboard mixture analysis and 24/7 reachback support reinforce operator confidence.
Ease of use for field operations
Designed for operators who work under pressure, Scan Assist technology guides users through sampling and technology selection, while customizable profiles assist the user with minimizing user error. Features like the motorized anvil and Scan Delay enhance safety by allowing operators to maintain distance from potential hazards. With intuitive workflows and a compact form factor, the Gemini™ analyzer reduces the burden of heavy equipment while increasing operational efficiency.
Add-on
Hazmaster G3: Integrated HazMasterG3® decision support software helps operators interpret results and assess potential threats from identified chemicals, supporting actionable insights directly in the field.
LowDoseID: By utilizing the enhanced hardware and software of the Gemini™ analyzer, combined with SERS, the Gemini™ analyzer with LowDoseID enables low concentration analysis and results for key narcotics.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Handheld Raman/FTIR Spectrometer
Certifications/Compliance Certified to MIL-STD-810G and IP67; full test report available on request
Collection Optics FTIR: ATR diamond crystal, single bounce, Raman: NA = 0.33
Data Export Formats SPC file (for use in standard spectroscopic software), text file, or JPEG report
Dimensions (L x W x H) 10.1 x 5.7 x 2.4 in. (25.6 x 14.6 x 6.1 cm)
Includes 5 years of support
IP Rating IP67
Languages English
Power Supply Internal 3.7 V Lithium Polymer Battery Pack ( QTY 1), Internal CR123A Batteries (QTY 3), DC Wall Adapter, 12 V DC, 1.25 A
Spectral Range FTIR: 4,000 cm-1 to 650 cm-1, Raman: 250 cm-1 to 2875 cm-1
Temperature Range (Metric) -20°C to +50°C
Weight (English) 4.2 lb.
Weight (Metric) 1.9 kg
Show More Show Less
What is Raman spectroscopy?
In Raman spectroscopy, an unknown sample of material is illuminated with monochromatic (single wavelength or single frequency) laser light, which can be absorbed, transmitted, reflected, or scattered by the sample. Light scattered from the sample is due to either elastic collisions of the light with the sample's molecules (Rayleigh scatter) or inelastic collisions (Raman scatter). Whereas Rayleigh scattered light has the same frequency (wavelength) of the incident laser light, Raman scattered light returns from the sample at different frequencies corresponding to the vibrational frequencies of the bonds of the molecules in the sample.
If you wish to learn more about Raman spectroscopy, visit our online Raman Spectroscopy Academy (https://www.thermofisher.com/us/en/home/industrial/spectroscopy-elemental-isotope-analysis/spectroscopy-elemental-isotope-analysis-learning-center/molecular-spectroscopy-information/raman-technology.html), where you will find basic Raman tutorials, advanced Raman webinars on sample applications, and a helpful instrument guide.
Do you offer support for the Gemini Raman and FTIR Chemical Analyzer with LowDoseID?
With an active support plan, Gemini products include 24/7, 365 reachback support with access to Ph.D. spectroscopists for spectral analysis and operational inquiries. Go here (https://portables.thermoscientific.com/gemini-customer-support-login) to access the Gemini customer support login page, where you can locate your Gemini serial number and support code, find contact information, access safety data sheets, ISO 9001 certificates, and warranty information, view parts and accessories, and log in to the download center.
How accurate is the Gemini Raman and FTIR Chemical Analyzer with LowDoseID?
The Gemini Raman and FTIR Chemical Analyzer with LowDoseID produces clear results, with no subjective interpretation. The instrument includes a comprehensive library of over 14,000 chemicals (both solids and liquids), and features best-in-class algorithms and automatic mixture analysis.
How easy is it to operate the Gemini Raman and FTIR Chemical Analyzer with LowDoseID?
The intuitive interface of the Gemini Raman and FTIR Chemical Analyzer with LowDoseID ensures quick training for first-time users and continued proficiency for experienced operators. The Scan Assist feature helps new users select the most appropriate technology with a few simple questions.
In addition, there is a large tactile keypad, a resistive touch screen designed for use while wearing gloves, an integrated Raman probe, motorized FTIR anvil, and flexible power options, among other user friendly features that cater to both experts and non-experts in the field.
How portable is the Gemini Raman and FTIR Chemical Analyzer with LowDoseID?
Weighing less than 2 kilograms, the Gemini Raman and FTIR Chemical Analyzer with LowDoseID is easily transported downrange or into the hazard zone, reducing the logistical burden of carrying two separate technologies and increasing efficiency with confirmatory identification capability when and where it is needed. By providing sophisticated technology in such a compact, lightweight device, one no longer needs to sacrifice capability for weight, giving an unparalleled advantage in the field.
Can the Gemini Raman and FTIR Chemical Analyzer with LowDoseID work in harsh conditions?
We understand the conditions operators face and the demands they place on equipment. From extreme climates to aggressive handling, the Gemini Raman and FTIR Chemical Analyzer with LowDoseID has been certified to the latest military standards for ruggedness - drop, shock, vibration, temperature, immersion, and much more.
How safe is the Gemini Raman and FTIR Chemical Analyzer with LowDoseID to operate?
In addition to Raman scan delay, adjustable laser power, and other built-in safety features, the Gemini Raman and FTIR Chemical Analyzer with LowDoseID includes the industry's first FTIR scan delay, enabled by a motorized anvil. Optional HazMaster G3 software enables users to virtually mix chemical substances, protecting themselves and the public from potentially dangerous chemical combinations.
Operators should take necessary precautions when operating the instrument, as outlined in the product user manual and training materials. Also, operators should ALWAYS follow their own organization's training and instructions, procedures, and regulations for the safe handling of unknown, hazardous, and/or energetic substances.
What technologies are included in the Gemini Raman and FTIR Chemical Analyzer with LowDoseID for chemical and explosives identification?
The Gemini Raman and FTIR Chemical Analyzer with LowDoseID enables operators to quickly switch between Raman and Fourier Transform Infrared (FTIR) spectroscopic analysis techniques. Raman and FTIR spectroscopy are highly specific and reliable identification methods, each with strengths and limitations. By integrating both into a single handheld analyzer, operators harness the power of each technology while enabling a broader range of chemical identification.