Learn More
DIATRON US Gemini Combo Automated ELISA & IFA Immunoassay System
Supplier:  DIATRON US 10041560
Encompass_Preferred
Gemini Combo is an Automated 96 Well ELISA System are computer-controlled microplate processing systems that fully automates ELISA assays. The Gemini line automates the sample distribution, incubation, reagent addition, washing, and detection phases of microplate assays. Gemini Combo can also fully process (prepared for reading) IFA slides in addition to being able to process 96-Well ELISA plates.
Catalog No. NC1847132
Only null left
Add to Cart
Request a Quote